Ammonium iodide ≥99.0% ACS

Supplier: BeanTown Chemical

139715-100G 139715-500G
BT139715-100GEA 76.53 USD
BT139715-100G BT139715-500G
Ammonium iodide ≥99.0% ACS
Ammonium iodide

CAS: 12027-06-4; EC No: 234-717-7; MDL No: MFCD00011424 Granular; Linear Formula: NH4I ; MW: 144.94 Melting Point: 551° (sublimes) Hygroscopic , Light Sensitive


  • Hygroscopic, Light Sensitive
  • Granular

Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551 (subl.)
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 13,00531

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR