Aluminium oxide ≥99% (trace metals basis) catalyst support, activated
Supplier: BeanTown Chemical
139650-1KG
139650-250G
BT139650-1KGEA
223
USD
BT139650-1KG
BT139650-250G
Aluminium oxide ≥99% (trace metals basis) catalyst support, activated
Aluminium oxide
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
3.2mm Pellets, S.A. 90 m2/g; Molecular Formula: Al2O3; MW: 101.96
Melting Point: 2040°
- 3.2mm Pellets, S.A. 90 m²/g
Formula:
Al₂O₃ Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...