Diammonium hydrogen citrate 98.0-103.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Ammonium citrate dibasic
, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate
, Citric acid diammonium salt
139645-500G
139645-2KG
139645-100G
BT139645-500GEA
86.28
USD
BT139645-500G
BT139645-2KG
BT139645-100G
Diammonium hydrogen citrate 98.0-103.0% ACS
di-Ammonium hydrogen citrate
- Granular
Formula:
HOC(CO₂H)(CH₂CO₂NH₄)₂ MW: 226.19 g/mol |
MDL Number:
MFCD00013068 CAS Number: 3012-65-5 EINECS: 221-146-3 |
Specification Test Results
Assay | 98.0-103.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...