Cadmium ≥99.995% (trace metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

139495-100X100 139495-50X50MM
BT139495-100X1EA 167 USD
BT139495-100X1 BT139495-50X50
Cadmium ≥99.995% (trace metals basis), foil, Thickness 0.25 mm
Cadmium

CAS: 7440-43-9; EC No: 231-152-8; MDL No: MFCD00010914 Solid; Molecular Formula: Cd; MW: 112.41 Melting Point: 320.9°; Boiling Point: 765° Density (g/mL): 8.65


  • Solid

Formula: Cd
MW: 112.41 g/mol
Boiling Pt: 765
Melting Pt: 321
Density: 8.65 g/mL
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1 II
Merck Index: 12,01649

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR