Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 63,000

Supplier: BeanTown Chemical

139370-1L 139370-250ML 139370-50ML
BT139370-1LEA 166 USD
BT139370-1L BT139370-250ML BT139370-50ML
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 63,000
Polydimethylsiloxane, trimethylsiloxy terminated

CAS: 9016-00-6; MDL No: MFCD00148360 Liquid; Linear Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 ; MW ≈63,000 Melting Point: -40° Density (g/mL): 0.974; Refractive Index: 1.404


  • Refractive Index: 1.404

Formula: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Melting Pt: –40 °C
Density: 0.974 g/ml
MDL Number: MFCD00148360
CAS Number: 9016-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR