Copper (I) sulfide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
139120-1KG
139120-250G
139120-50G
BT139120-1KGEA
879
USD
BT139120-1KG
BT139120-250G
BT139120-50G
Copper (I) sulfide ≥99.5% (trace metals basis)
Copper (I) sulphide
CAS: 22205-45-4; EC No: 244-842-9; MDL No: MFCD00016062
-200 Mesh Powder; Molecular Formula: Cu2S; MW: 159.15
Melting Point: 1100°
Density (g/mL): 5.6
- -200 Mesh Powder
Formula:
Cu₂S Melting Pt: 1100 °C Density: 5.6 g/mL |
MDL Number:
MFCD00016062 CAS Number: 22205-45-4 EINECS: 244-842-9 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...