Polyethyleneimine, M.W. ∼600 ≥99%, branched

Supplier: BeanTown Chemical

Synonyms: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)

139110-100G 139110-25G 139110-500G
BT139110-100GEA 142.06 USD
BT139110-100G BT139110-25G BT139110-500G
Polyethyleneimine, M.W. ∼600 ≥99%, branched
Polyethyleneimine

CAS: 9002-98-6; MDL No: MFCD00803910 Liquid; Linear Formula: (-NHCH2CH2-)x[-N(CH2CH2NH2)CH2CH2-]y


  • Liquid

Formula: (CH2CH2NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR