Tetrathiafulvalene ≥97%

Supplier: BeanTown Chemical

139105-1G 139105-250MG
BT139105-1GEA 169 USD
BT139105-1G BT139105-250MG
Tetrathiafulvalene ≥97%
Tetrathiafulvalene

CAS: 31366-25-3; EC No: 250-593-7; MDL No: MFCD00005492 Crystalline; Molecular Formula: C6H4S4; MW: 204.35 Melting Point: 116-119° Air Sensitive, Light Sensitive


  • Air Sensitive, Light Sensitive
  • Crystalline

Formula: C₆H₄S₄
Melting Pt: 116…119 °C
MDL Number: MFCD00005492
CAS Number: 31366-25-3
EINECS: 250-593-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR