Tin ≥99.9% (trace metals basis), rods Ø 12.7 mm

Supplier: BeanTown Chemical

138965-100CM 138965-25CM
BT138965-100CMEA 244 USD
BT138965-100CM BT138965-25CM
Tin ≥99.9% (trace metals basis), rods Ø 12.7 mm
Tin

CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862 Solid; Molecular Formula: Sn; MW: 118.71 Melting Point: 231.9°; Boiling Point: 2270° Density (g/mL): 7.31


  • Solid

Formula: Sn
Boiling Pt: 2270 °C
Melting Pt: 231.9 °C
Density: 7.31 g/mL
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR