Calcium carbonate ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Marble

138870-100G 138870-2KG 138870-500G
BT138870-100GEA 56.51 USD
BT138870-100G BT138870-2KG BT138870-500G
Calcium carbonate ≥99.5%
Calcium carbonate

CAS: 471-34-1; EC No: 207-439-9; MDL No: MFCD00010906; RTECS: FF9335000 5 Micron Powder; Linear Formula: CaCO3; MW: 100.09 Density (g/mL): 2.93


  • RTECS: FF9335000
  • 5 Micron Powder

Formula: CaCO₃
Density: 2.93 g/mL
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR