Tellurium ≥99.999% (trace metals basis), lumps

Supplier: BeanTown Chemical

138440-100G 138440-25G 138440-500G
BT138440-100GEA 145.05 USD
BT138440-100G BT138440-25G BT138440-500G
Tellurium ≥99.999% (trace metals basis), lumps
Tellurium

CAS: 13494-80-9; EC No: 236-813-4; MDL No: MFCD00134062; RTECS: WY2625000 Solid; Molecular Formula: Te; MW: 127.60 Melting Point: 450°; Boiling Point: 990° Density (g/mL): 6.24


  • RTECS: WY2625000
  • Solid

Formula: Te
Boiling Pt: 990 °C
Melting Pt: 450 °C
Density: 6.24 g/mL
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR