Disodium oxalate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
137765-100G
137765-2.5KG
137765-500G
BT137765-100GEA
54.96
USD
BT137765-100G
BT137765-2.5KG
BT137765-500G
Disodium oxalate ≥99%
di-Sodium oxalate
CAS: 62-76-0; EC No: 200-550-3; MDL No: MFCD00012465; RTECS: KI1750000
Solid; Linear Formula: Na2C2O4; MW: 134.00
Melting Point: 250-270°
Density (g/mL): 2.34
Hygroscopic
- RTECS: KI1750000
- Hygroscopic
- Solid
Formula:
Na₂C₂O₄ Melting Pt: 250…270 °C Density: 2.34 g/mL |
MDL Number:
MFCD00012465 CAS Number: 62-76-0 EINECS: 200-550-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...