1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97%

Supplier: BeanTown Chemical

Synonyms: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;

137180-25G 137180-5G
BT137180-25GEA 309.95 USD
BT137180-25G BT137180-5G
1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97%
1H,1H,2H,2H-Perfluorooctyltriethoxysilane

CAS: 51851-37-7; EC No: 257-473-3; MDL No: MFCD00042333 Liquid; Molecular Formula: C14H19F13O3Si; MW: 510.36 Flash point: 97°C (207°F) Density (g/mL): 1.33; Refractive Index: 1.344 Moisture Sensitive


  • Refractive index: 1.344
  • Moisture Sensitive
  • Liquid

Formula: C₁₄H₁₉F₁₃O₃Si
MW: 510.367 g/mol
Density: 1.33 g/mL
Flash Pt: 97 °C (207 °F)
MDL Number: MFCD00042333
CAS Number: 51851-37-7
EINECS: 257-473-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR