Diammonium-L(+)-tartrate ≥98%

Supplier: BeanTown Chemical

Synonyms: (R,R)-(+)-Diammonium tartrate, (R,R)-(+)-Ammonium tartrate dibasic, Ammonium tartrate dibasic, L(+)-Tartaric acid diammonium salt, (+)-Diammonium L-tartrate, (R,R)-(+)-Tartaric acid diammonium salt, L(+)-Ammonium tartrate dibasic, Ammonium L-(+)-tartrate

137000-1KG 137000-250G 137000-50G
BT137000-1KGEA 153 USD
BT137000-1KG BT137000-250G BT137000-50G
Diammonium-L(+)-tartrate ≥98%
di-Ammonium L(+)-tartrate

CAS: 3164-29-2; EC No: 221-618-9; MDL No: MFCD00013073; RTECS: WW8050000 Crystalline; Molecular Formula: C4H12N2O6; MW: 184.15 Hygroscopic


  • RTECS: WW8050000
  • Hygroscopic
  • Crystalline

Formula: C₄H₁₂N₂O₆
MW: 184.15 g/mol
MDL Number: MFCD00013073
CAS Number: 3164-29-2
EINECS: 221-618-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR