Silica gel, Pore size: 60 Å 0.036-0.071 mm (215-400 mesh)

Supplier: BeanTown Chemical

136860-1KG 136860-2.5KG
BT136860-1KGEA 116.47 USD
BT136860-1KG BT136860-2.5KG
Silica gel, Pore size: 60 Å 0.036-0.071 mm (215-400 mesh)
Silica gel

CAS: 63231-67-4; RTECS: VV7565000 Powder Melting Point: 1600°; Boiling Point: 2230° Hygroscopic


  • RTECS: VV7565000
  • Hygroscopic
  • Powder

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: >1600 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR