Anethole trithione ≥95%

Supplier: BeanTown Chemical

Synonyms: 5-(4-Methoxyphenyl)-3H-1,2-dithiole-3-thione, Felviten

136610-1G 136610-5G
BT136610-1GEA 135 USD
BT136610-1G BT136610-5G
Anethole trithione ≥95%
Anethole trithione

CAS: 532-11-6; EC No: 208-528-5; MDL No: MFCD00129751 Solid; Molecular Formula: C10H8OS3; MW: 240.37


  • Solid

Formula: C₁₀H₈OS₃
MDL Number: MFCD00129751
CAS Number: 532-11-6
EINECS: 208-528-5

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR