Samarium ≥99.9% (trace rare earth metals basis), ingot
Supplier: BeanTown Chemical
136115-100G
136115-25G
BT136115-100GEA
286
USD
BT136115-100G
BT136115-25G
Samarium ≥99.9% (trace rare earth metals basis), ingot
Samarium
CAS: 7440-19-9; EC No: 231-128-7; MDL No: MFCD00011233
Solid, packaged in air stable poly bottle; Molecular Formula: Sm; MW: 150.36
Melting Point: 1074°; Boiling Point: 1794°
Density (g/mL): 7.47
- Solid, packaged in air stable poly bottle
Formula:
Sm Boiling Pt: 1794 °C Melting Pt: 1074 °C Density: 7.47 g/mL |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...