Tin(IV) oxide ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Tin(iv)oxide, Stannic Oxide, Tin dioxide
136065-25G
136065-5G
BT136065-25GEA
460
USD
BT136065-25G
BT136065-5G
Tin(IV) oxide ≥99.995%
Tin (IV) oxide
CAS: 18282-10-5; EC No: 242-159-0; MDL No: MFCD00011244; RTECS: XQ4000000
-22 Mesh Powder; Linear Formula: SnO2; MW: 150.71
Melting Point: 1630°; Boiling Point: 1800-1900°
Density (g/mL): 6.95; Refractive Index: 1.997
- RTECS: XQ4000000
- Refractive index: 1.997
- -22 Mesh Powder
Formula:
O₂Sn MW: 150.71 g/mol Boiling Pt: 1800…1900 °C Melting Pt: 1624.85 Density: 6.95 g/mL |
MDL Number:
MFCD00011244 CAS Number: 18282-10-5 EINECS: 242-159-0 Merck Index: 14,08778 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...