Butyl-4-hydroxybenzoate 99

Supplier: BeanTown Chemical

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

136020-1KG 136020-250G 136020-5KG
BT136020-1KGEA 183 USD
BT136020-1KG BT136020-250G BT136020-5KG
Butyl-4-hydroxybenzoate 99
Butyl 4-hydroxybenzoate

CAS: 94-26-8; EC No: 202-318-7; MDL No: MFCD00016478; RTECS: DH1980000 Powder; Molecular Formula: C11H14O3 ; MW: 194.23 Melting Point: 67-70°; Boiling Point: 156-157°/3.5 mmHg


  • RTECS: DH1980000
  • Powder

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156…157 °C/3.5 mmHg
Melting Pt: 73
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 13,01583

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR