Tantalum ≥99.9% (trace metals basis), plate, Thickness 6.35 mm

Supplier: BeanTown Chemical

136005-25X25MM 136005-50X50MM
BT136005-25X25EA 577 USD
BT136005-25X25 BT136005-50X50
Tantalum ≥99.9% (trace metals basis), plate, Thickness 6.35 mm
Tantalum

CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252; RTECS: WW5505000 Solid; Molecular Formula: Ta; MW: 180.95 Melting Point: 2996°; Boiling Point: 5425° Density (g/mL): 16.69


  • RTECS: WW5505000
  • Solid

Formula: Ta
Boiling Pt: 5425 °C
Melting Pt: 2996 °C
Density: 16.69 g/mL
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR