Graphite (natural) ≥99.9% (trace metals basis), soluble -10 mesh

Supplier: BeanTown Chemical

135915-1KG 135915-250G 135915-50G
BT135915-1KGEA 81 USD
BT135915-1KG BT135915-250G BT135915-50G
Graphite (natural) ≥99.9% (trace metals basis), soluble -10 mesh
Graphite

CAS: 7782-42-5; EC No: 231-955-3; MDL No: MFCD00144065 Solid; Molecular Formula: C; MW: 12.011


  • Solid

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR