Charcoal activated -20+40 mesh

Supplier: BeanTown Chemical

Synonyms: Carbon activated, Activated carbon, Activated charcoal

135855-100G 135855-2KG 135855-500G
BT135855-100GEA 22.05 USD
BT135855-100G BT135855-2KG BT135855-500G
Charcoal activated -20+40 mesh
Charcoal activated

CAS: 7440-44-0; EC No: 231-153-3; MDL No: MFCD00133992 Powder; Molecular Formula: C; MW: 12.01 Melting Point: 3550°


  • Powder

Formula: C
Melting Pt: 3550 °C
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR