Indium ≥99.99% (trace metals basis), tear drop shot 4 mm
Supplier: BeanTown Chemical
135830-10G
135830-250G
135830-50G
BT135830-10GEA
88.47
USD
BT135830-10G
BT135830-250G
BT135830-50G
Indium ≥99.99% (trace metals basis), tear drop shot 4 mm
Indium
CAS: 7440-74-6; EC No: 231-180-0; MDL No: MFCD00134048; RTECS: NL1050000
Solid; Molecular Formula: In; MW: 114.82
Melting Point: 156.6°; Boiling Point: 2072°
Density (g/mL): 7.3
- RTECS: NL1050000
- Solid
Formula:
In Boiling Pt: 2072 °C Melting Pt: 156.6 °C Density: 7.3 g/mL |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...