Dimethyl sulfoxide-[D₆] ≥100% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulfoxide, Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulfoxide-[²H₆], Dimethyl sulphoxide-d6, Dimethyl sulfoxide-d6
135815-10EACH
135815-2EACH
BT135815-10EACEA
307
USD
BT135815-10EAC
BT135815-2EACH
Dimethyl sulfoxide-[D₆] ≥100% (Isotopic)
Dimethyl sulphoxide-[D6]
CAS: 2206-27-1; EC No: 218-617-0; MDL No: MFCD00002090
Liquid, in prescored ampules, 0.75ml/ampoule ; Linear Formula: (CD3)2SO; MW: 84.17
Melting Point: 20°; Boiling Point: 189°; Flash point: 87°C (189°F)
Density (g/mL): 1.19; Refractive Index: 1.476
Hygroscopic
- Refractive index: 1.476
- Hygroscopic
- Liquid, in prescored ampules, 0.75ml/ampoule
Formula:
D₃CS(O)CD₃ MW: 84.09 g/mol Boiling Pt: 189 Melting Pt: 20.2 Density: 1.19 g/mL Flash Pt: 87 °C (189 °F) |
MDL Number:
MFCD00002090 CAS Number: 2206-27-1 EINECS: 218-617-0 |
Specification Test Results
Assay | 100% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...