Calcium fluoride ≥99.5%

Supplier: BeanTown Chemical

135790-1KG 135790-250G 135790-5KG
BT135790-1KGEA 181 USD
BT135790-1KG BT135790-250G BT135790-5KG
Calcium fluoride ≥99.5%
Calcium fluoride

CAS: 7789-75-5; EC No: 232-188-7; MDL No: MFCD00010907; RTECS: EW1760000 Powder; Molecular Formula: CaF2; MW: 78.08 Boiling Point: 2500° Density (g/mL): 3.18 Hygroscopic


  • RTECS: EW1760000
  • Hygroscopic
  • Powder

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500
Melting Pt: 1418
Density: 3.18 g/mL
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR