Aluminum metaphosphate
Supplier: BeanTown Chemical
135050-1KG
135050-250G
BT135050-1KGEA
94
USD
BT135050-1KG
BT135050-250G
Aluminum metaphosphate
Aluminum metaphosphate
CAS: 32823-06-6; MDL No: MFCD00075314
Powder; Linear Formula: Al(PO3)3 ; MW: 263.90
Density (g/mL): 2.779
- Powder
Formula:
Al(PO₃)₃ Density: 2.779 g/mL |
MDL Number:
MFCD00075314 CAS Number: 32823-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...