4-Nitrophthalhydrazide ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Nitrophthalhydrazide, 2,3-Dihydro-6-nitro-1,4-phthalazinedione, 4-Nitrophthalic Hydrazide
134335-25G
134335-5G
BT134335-25GEA
175
USD
BT134335-25G
BT134335-5G
4-Nitrophthalhydrazide ≥98%
4-Nitrophthalhydrazide
CAS: 3682-19-7; MDL No: MFCD00006891; RTECS: TH8895000
Powder; Molecular Formula: C8H5N3O4; MW: 207.14
- RTECS: TH8895000
- Powder
Formula:
C₈H₅N₃O₄ |
MDL Number:
MFCD00006891 CAS Number: 3682-19-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...