Tartrazine

Supplier: BeanTown Chemical

Synonyms: FD and C Yellow No. 5Sodium 5-oxo-1-(4-sulphonatophenyl)-4-((4-sulphonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylateAcid Yellow 23Sodium 5-oxo-1-(4-sulfonatophenyl)-4-((4-sulfonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylateFilter Yellow

134190-25G 134190-100G 134190-500G
BT134190-25GEA 26.21 USD
BT134190-25G BT134190-100G BT134190-500G
Tartrazine
Tartrazine

  • RTECS: UQ6400000
  • Hygroscopic
  • Solid

Formula: C₁₆H₁₂N₄O₉S₂
MW: 534.36 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00148908
CAS Number: 1934-21-0
EINECS: 217-699-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR