Aluminium ≥99% (trace metals basis), foil, not supported, Thickness 0.8 µm

Supplier: BeanTown Chemical

Synonyms: Aluminum

134150-140X140 134150-160X160 134150-30X30MM
BT134150-140X1EA 207 USD
BT134150-140X1 BT134150-160X1 BT134150-30X30
Aluminium ≥99% (trace metals basis), foil, not supported, Thickness 0.8 µm
Aluminium

CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029 Solid; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7


  • Solid

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR