Potassium carbonate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Potash, Carbonic acid potassium salt, Carbonic acid, dipotassium salt

133715-100G 133715-25G 133715-500G
BT133715-100GEA 54.83 USD
BT133715-100G BT133715-25G BT133715-500G
Potassium carbonate ≥99.0% ACS
Potassium carbonate

CAS: 584-08-7; EC No: 209-529-3; MDL No: MFCD00011382; RTECS: TS7750000 Granular; Linear Formula: K2CO3; MW: 138.21 Melting Point: 891° Density (g/mL): 2.43 Hygroscopic


  • RTECS: TS7750000
  • Hygroscopic
  • Granular

Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Density: 2.43 g/mL
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.
Insoluble matter ≤0.01%
Nitrogen compounds (as N) ≤0.001%
Chloride (Cl) ≤0.003%
Phosphate (PO4) ≤0.001%
Heavy metals ≤5ppm
Ammonium hydroxide PPT ≤0.01%
Iron (Fe) ≤5ppm
Arsenic (As) ≤1ppm
Sodium (Na) ≤0.02%
Sulfur Compounds ≤0.003%
Silica (SiO2) ≤0.005%
Magnesium (Mg) ≤0.002%
Calcium (Ca) ≤0.005%
Appearance While small granules
Clarity of solution Clear and complete


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR