Calcium molybdate ≥99.5%

Supplier: BeanTown Chemical

133670-100G 133670-25G
BT133670-100GEA 149 USD
BT133670-100G BT133670-25G
Calcium molybdate ≥99.5%
Calcium molybdate

CAS: 7789-82-4; EC No: 232-192-9; MDL No: MFCD00015976; RTECS: EW2975000 Powder; Molecular Formula: CaMoO4 ; MW: 200.01


  • RTECS: EW2975000
  • Powder

Formula: CaMoO₄
MW: 200.024 g/mol
MDL Number: MFCD00015976
CAS Number: 7789-82-4
EINECS: 232-192-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR