Lactic acid 85,0-90,0 w/w water ACS

Supplier: BeanTown Chemical

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

133595-100G 133595-2KG 133595-500G
BT133595-100GEA 32.63 USD
BT133595-100G BT133595-2KG BT133595-500G
Lactic acid 85,0-90,0 w/w water ACS
Lactic acid

CAS: 50-21-5; EC No: 200-018-0; MDL No: MFCD00004520; RTECS: OD2800000 Liquid; Molecular Formula: C3H6O3; MW: 90.08 Flash point: 110°C (230°F) Density (g/mL): 1.209; Refractive Index: 1.425


  • RTECS: OD2800000
  • Refractive index: 1.425
  • Liquid

Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.209 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
Merck Index: 13,05351

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR