Niobium(V) oxide ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Diniobium pentaoxide, Niobium pentoxide

133550-1KG 133550-250G 133550-50G
BT133550-1KGEA 1396 USD
BT133550-1KG BT133550-250G BT133550-50G
Niobium(V) oxide ≥99.995%
Niobium (V) oxide

CAS: 1313-96-8; EC No: 215-213-6; MDL No: MFCD00011128; RTECS: QU0500000 Powder; Molecular Formula: Nb2O5; MW: 265.81 Melting Point: 1520° Density (g/mL): 4.47


  • RTECS: QU0500000
  • Powder

Formula: Nb₂O₅
MW: 265.81 g/mol
Melting Pt: 1496
Density: 4.47 g/mL
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR