Dimethyl sulfoxide-[D₆] ≥99.9% (Isotopic)

Supplier: BeanTown Chemical

133370-10X1G 133370-5X1G
BT133370-10X1GEA 101.47 USD
BT133370-10X1G BT133370-5X1G
Dimethyl sulfoxide-[D₆] ≥99.9% (Isotopic)
Dimethyl sulphoxide-[D6]

CAS: 2206-27-1; EC No: 218-617-0; MDL No: MFCD00002090 Liquid; Linear Formula: (CD3)2SO; MW: 84.17 Melting Point: 20°; Boiling Point: 189°; Flash point: 87°C (189°F) Density (g/mL): 1.19; Refractive Index: 1.476 Hygroscopic


  • Refractive index: 1.476
  • Hygroscopic
  • Liquid

Formula: D₃CS(O)CD₃
Boiling Pt: 189 °C
Melting Pt: 20 °C
Density: 1.19 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR