Molecular sieve A4 (0.4 nm, 4 Å), beads 1 - 2 mm
Supplier: BeanTown Chemical
Synonyms:
Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
133105-1KG
133105-250G
133105-5KG
BT133105-1KGEA
124.29
USD
BT133105-1KG
BT133105-250G
BT133105-5KG
Molecular sieve A4 (0.4 nm, 4 Å), beads 1 - 2 mm
Molecular sieve A4 (0.4 nm, 4 Å)
CAS: 70955-01-0; MDL No: MFCD00131613
Beads; Linear Formula: Na12[(AlO2)12(SiO2)12]·xH2O
Hygroscopic
- Hygroscopic
- Beads
MDL Number:
MFCD03457537 CAS Number: 70955-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...