Iodine 0.01 N water Volumetric

Supplier: BeanTown Chemical

132950-1L 132950-4L
BT132950-1LEA 41.23 USD
BT132950-1L BT132950-4L
Iodine 0.01 N water Volumetric
Iodine

CAS: 7553-56-2; EC No: 231-442-4; MDL No: MFCD00011355 Liquid; Molecular Formula: I2; MW: 253.81


  • Liquid

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3264
ADR: 8 II
Merck Index: 13,05036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR