m-Cresol purple sodium salt

Supplier: BeanTown Chemical

Synonyms: m-Cresolsulfonphthalein sodium salt, m-Cresolsulphonphthalein sodium salt, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[d][1,2]oxathiole 2,2-dioxide, sodium salt

132900-25G 132900-5G
BT132900-25GEA 177 USD
BT132900-25G BT132900-5G
m-Cresol purple sodium salt
m-Cresol purple sodium salt

CAS: 62625-31-4; EC No: 263-656-9; MDL No: MFCD00010177 Powder; Molecular Formula: C21H17NaO5S; MW: 404.42


  • Powder

Formula: C₂₁H₁₇NaO₅S
MW: 404.42 g/mol
MDL Number: MFCD00010177
CAS Number: 62625-31-4
EINECS: 263-656-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR