Ethylene glycol ≥99%

Supplier: BeanTown Chemical

132725-2.5KG 132725-500G
BT132725-2.5KGEA 90.11 USD
BT132725-2.5KG BT132725-500G
Ethylene glycol ≥99%
Ethylene glycol

CAS: 107-21-1; EC No: 203-473-3; MDL No: MFCD00002885; RTECS: KW2975000 Liquid; Linear Formula: HOCH2CH2OH; MW: 62.07 Melting Point: -13°; Boiling Point: 196-198°; Flash point: 119°C (246°F) Density (g/mL): 1.113; Refractive Index: 1.431


  • RTECS: KW2975000
  • Refractive index: 1.431
  • Liquid

Formula: HOCH₂CH₂OH
Boiling Pt: 196…198 °C
Melting Pt: –13 °C
Density: 1.113 g/mL
Flash Pt: 119 °C (246 °F)
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR