Sulfanilic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

132640-1KG 132640-250G 132640-5KG
BT132640-1KGEA 57.08 USD
BT132640-1KG BT132640-250G BT132640-5KG
Sulfanilic acid ≥98%
Sulphanilic acid

CAS: 121-57-3; EC No: 204-482-5; MDL No: MFCD00007886; RTECS: WP3895500 Crystalline/Pieces; Linear Formula: H2NC6H4SO3H; Molecular Formula: C6H7NO3S; MW: 173.19 Melting Point: 365° Density (g/mL): 1.485


  • RTECS: WP3895500
  • Crystalline/Pieces

Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.485 g/mL
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR