Ammonium hexachloroosmate(IV) ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Diammonium hexachloroosmate
132440-1G
132440-5G
BT132440-1GEA
111
USD
BT132440-1G
BT132440-5G
Ammonium hexachloroosmate(IV) ≥99.9%
Ammonium hexachloroosmate(IV)
CAS: 12125-08-5; EC No: 235-188-5; MDL No: MFCD00010883
Crystalline; Linear Formula: (NH4)2OsCl6; MW: 439.02
Melting Point: 170° (sublimes)
Density (g/mL): 2.93
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
H₈Cl₆N₂Os MW: 439.02 g/mol Melting Pt: 170 (subl.) Density: 2.93 g/mL |
MDL Number:
MFCD00010883 CAS Number: 12125-08-5 EINECS: 235-188-5 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...