Sodium alginate
Supplier: BeanTown Chemical
Synonyms:
Alginic acid sodium salt, Algin
132270-100G
132270-250G
BT132270-100GEA
70.07
USD
BT132270-100G
BT132270-250G
Sodium alginate
Sodium alginate
CAS: 9005-38-3; MDL No: MFCD00081310; RTECS: AZ5820000
Powder
MDL Number:
MFCD00081310 CAS Number: 9005-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...