di-Sodium succinate hexahydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium succinate dibasic hexahydrate
, Succinic acid disodium salt hexahydrate
, Sodium succinate hexahydrate
, Succinic acid disodium hexahydrate
, Sodium Succinate, Hexahydrate
, Disodium succinate hexahydrate
, 1,4-Butanedioic acid disodium salt hexahydrate
132110-250G
132110-1KG
132110-5KG
BT132110-250GEA
52.45
USD
BT132110-250G
BT132110-1KG
BT132110-5KG
di-Sodium succinate hexahydrate ≥99%
di-Sodium succinate hexahydrate
- RTECS: WM7751000
- Hygroscopic
- Crystalline
Formula:
NaOOCCH₂CH₂COONa·6H₂O MW: 270.14 g/mol |
MDL Number:
MFCD00149117 CAS Number: 6106-21-4 EINECS: 205-778-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...