3,4,9,10-Perylenetetracarboxylic dianhydride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA
132070-10G
132070-50G
BT132070-10GEA
29
USD
BT132070-10G
BT132070-50G
3,4,9,10-Perylenetetracarboxylic dianhydride ≥98%
3,4,9,10-Perylenetetracarboxylic dianhydride
CAS: 128-69-8; EC No: 204-905-3; MDL No: MFCD00006916
Powder; Molecular Formula: C24H8O6; MW: 392.32
Melting Point:
- Moisture Sensitive
- Powder
Formula:
C₂₄H₈O₆ MW: 392.32 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00006916 CAS Number: 128-69-8 EINECS: 204-905-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...