Silica gel, Pore size: 150 Å 100-200 mesh, wide pore
Supplier: BeanTown Chemical
132060-1KG
132060-250G
132060-50G
BT132060-1KGEA
375.86
USD
BT132060-1KG
BT132060-250G
BT132060-50G
Silica gel, Pore size: 150 Å 100-200 mesh, wide pore
Silica gel
CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232; RTECS: VV7565000
Powder, S.A. 350-400 m2/g
Melting Point:
- RTECS: VV7565000
- Hygroscopic
- Powder, S.A. 350-400 m²/g
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: >1600 °C |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...