Anthrone ACS
Supplier: BeanTown Chemical
Synonyms:
Anthracen-9(10H)-one
131945-100G
131945-25G
BT131945-100GEA
173.62
USD
BT131945-100G
BT131945-25G
Anthrone ACS
Anthrone
CAS: 90-44-8; EC No: 201-994-0; MDL No: MFCD00001187; RTECS: CB8925500
Crystalline; Molecular Formula: C14H10O; MW: 194.23
Melting Point: 154-157°
Light Sensitive
- RTECS: CB8925500
- Light Sensitive
- Crystalline
Formula:
C₁₄H₁₀O MW: 194.23 g/mol Melting Pt: 154…157 °C |
MDL Number:
MFCD00001187 CAS Number: 90-44-8 EINECS: 201-994-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...