Silicon ≥99.9999% (trace metals basis), lumps 0.1-2.5cm

Supplier: BeanTown Chemical

131940-100G 131940-500G
BT131940-100GEA 147.2 USD
BT131940-100G BT131940-500G
Silicon ≥99.9999% (trace metals basis), lumps 0.1-2.5cm
Silicon

CAS: 7440-21-3; EC No: 231-130-8; MDL No: MFCD00085311; RTECS: VW0400000 Solid; Molecular Formula: Si; MW: 28.09 Melting Point: 1410°; Boiling Point: 2355° Density (g/mL): 2.33


  • RTECS: VW0400000
  • Solid

Formula: Si
Boiling Pt: 2355 °C
Melting Pt: 1410 °C
Density: 2.33 g/mL
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8

Order Now

Specification Test Results

Assay 99.9999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR