D-(-)-Mannitol 99

Supplier: BeanTown Chemical

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

131640-1KG 131640-250G 131640-5KG
BT131640-1KGEA 77.23 USD
BT131640-1KG BT131640-250G BT131640-5KG
D-(-)-Mannitol 99
D(-)-Mannitol

CAS: 69-65-8; EC No: 200-711-8; MDL No: MFCD00064287; RTECS: OP2060000 Powder; Molecular Formula: C6H14O6; MW: 182.17 Melting Point: 167-170°


  • RTECS: OP2060000
  • Powder

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Melting Pt: 167…170 °C
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR