Dowex® 50WX2 (H) 100-200 (H)

Supplier: BeanTown Chemical

131090-50G
BT131090-50GEA 59.31 USD
BT131090-50G
Dowex® 50WX2 (H) 100-200 (H)
Dowex® 50WX2 (H)

  • Beads

MDL Number: MFCD00132722
CAS Number: 12612-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR