Silica gel, Pore size: 60 Å 0.032-0.063mm; 230-450 mesh

Supplier: BeanTown Chemical

130930-1KG 130930-2.5KG 130930-500G
BT130930-1KGEA 124.9 USD
BT130930-1KG BT130930-2.5KG BT130930-500G
Silica gel, Pore size: 60 Å 0.032-0.063mm; 230-450 mesh
Silica gel

CAS: 63231-67-4; RTECS: VV7565000 Powder Melting Point: 1600°; Boiling Point: 2230° Hygroscopic


  • RTECS: VV7565000
  • Hygroscopic
  • Powder

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: >1600 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR