Vanadium ≥99.5% (trace metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

130860-150X200 130860-50X150
BT130860-150X2EA 578 USD
BT130860-150X2 BT130860-50X15
Vanadium ≥99.5% (trace metals basis), foil, Thickness 0.25 mm
Vanadium

CAS: 7440-62-2; EC No: 231-171-1; MDL No: MFCD00011453 Solid; Molecular Formula: V; MW: 50.94 Melting Point: 1890°; Boiling Point: 3380° Density (g/mL): 6.11


  • Solid

Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.11 g/mL
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR